Assay Details
Gene Symbol: KDELC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153705.4 | NP_714916.3 | 4 - 5 | 969 | 84 | |
XM_011542621.2 | XP_011540923.1 | 4 - 5 | 942 | 84 | ||
GenBank mRNA | AF533708.1 | - | 4 - 5 | 928 | 84 | |
AK125519.1 | - | 3 - 4 | 751 | 84 | ||
AK313534.1 | - | 4 - 5 | 831 | 84 | ||
AY358616.1 | - | 3 - 4 | 1038 | 84 | ||
BC036526.2 | - | 4 - 5 | 923 | 5266665 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002268.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027184.hg.1 | ENST00000323468.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027184.hg.1 | ENST00000375648.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027184.hg.1 | ENST00000434945.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027184.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027184.hg.1 | TC11002268.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194083.hg.1 | TC1100012229.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000524787.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | ENST00000532730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012229.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000323468.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000434945.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000530318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | ENST00000530529.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | NM_153705.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194082.hg.1 | TC1100012229.hg.1 | Clariom D TC |
