Assay Details
| Gene Symbol: BDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170732.4 | NP_733928.1 | 1 - 2 | 440 | 150 | |
| GenBank mRNA | AB038671.1 | - | 1 - 2 | 340 | 150 | |
| AK289763.1 | - | 1 - 2 | 139 | 150 | ||
| AY054395.2 | - | 1 - 2 | 384 | 150 | ||
| EF674519.1 | - | 1 - 2 | 440 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB038671.1, AK289763.1, AY054395.2, EF674519.1, NM_170732.4 transcripts.
Set Membership: |
|

BDNF
Genomic Map