Assay Details
Gene Symbol: LRRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321961.1 | NP_001308890.1 | 1 - 2 | 133 | 112 | |
NM_012472.5 | NP_036604.2 | 1 - 2 | 133 | 112 | ||
NR_073525.2 | 1 - 2 | 133 | 112 | |||
NR_135905.1 | 1 - 2 | 133 | 112 | |||
NR_135907.1 | 1 - 2 | 133 | 112 | |||
XM_017013298.1 | XP_016868787.1 | 1 - 2 | 74 | 112 | ||
GenBank mRNA | AK313107.1 | - | 1 - 2 | 108 | 112 | |
BC027589.1 | - | 1 - 2 | 79 | 4831028 | 112 | |
BC047286.1 | - | 1 - 2 | 83 | 5168914 | 112 | |
DR002710.1 | - | 1 - 2 | 119 | 112 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001654.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC027589.1,BC047286.1,NM_001321961.1,NM_012472.5,XM_017013298.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000250173.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518101.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000518642.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000519595.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000520446.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000521430.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522584.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522597.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000522789.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000523503.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000618342.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | ENST00000620350.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NM_012472.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800011861.hg.1 | NR_073525.2 | Refseq |
