Assay Details
| Gene Symbol: SVOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018711.4 | NP_061181.1 | 12 - 13 | 1530 | 82 | |
| GenBank mRNA | AK309780.1 | - | 3 - 4 | 275 | 82 | |
| BC033587.1 | - | 12 - 13 | 1330 | 5243017 | 82 | |
| BC094722.1 | - | 12 - 13 | 1177 | 4817829 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191936.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191936.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191936.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191936.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191936.hg.1 | TC1200011855.hg.1 | Clariom D TC |

SVOP
Genomic Map