Assay Details
| Gene Symbol: SVOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018711.4 | NP_061181.1 | 14 - 15 | 1720 | 119 | |
| GenBank mRNA | AK309780.1 | - | 5 - 6 | 465 | 119 | |
| AL359592.1 | - | 2 - 3 | 138 | 119 | ||
| BC033587.1 | - | 14 - 15 | 1520 | 5243017 | 119 | |
| BC094722.1 | - | 14 - 15 | 1367 | 4817829 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191934.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191934.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191934.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200191934.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200191934.hg.1 | TC1200011855.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000546618.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000548229.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000550436.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000551683.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000569754.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | ENST00000610966.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011855.hg.1 | NM_018711.4 | Refseq |

SVOP
Genomic Map