Assay Details
| Gene Symbol: RBM48 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032120.3 | NP_115496.2 | 5 | 3126 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864639 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864637 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05613519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864642 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864640 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864641 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864638 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P05613517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P25864643 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 440203 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P59714 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P4036 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P59714 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 29501 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 21491 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P112322 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P112322 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 4075 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P59714 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P59714 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15988919 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864607 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864620 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864611 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864620 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864608 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613505 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864624 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864607 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613495 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864616 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05613496 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15988919 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25864611 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P59714 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P4036 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P112322 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P59714 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P4036 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 30913 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 11742 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01754976 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613510 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988903 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613516 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613495 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613462 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988902 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613518 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988919 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988901 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613505 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613515 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613470 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613497 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988921 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613456 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613521 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613524 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613498 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613527 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613500 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613492 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613459 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15988898 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613523 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613469 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P05613507 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 1553881_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553881_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553881_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553881_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034709 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A034709 | - | - |

RBM48
Genomic Map