Assay Details
| Gene Symbol: CXCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032966.2 | NP_116743.1 | 1 | 160 | 114 | |
| GenBank mRNA | X68829.1 | - | 1 | 160 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001073.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_032966.2,X68829.1 transcripts.
Set Membership: |
|

CXCR5
Genomic Map