Assay Details
| Gene Symbol: DMRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527214.1 | XP_011525516.1 | 4 - 5 | 500 | 88 | |
| XM_011527215.1 | XP_011525517.1 | 4 - 5 | 492 | 88 | ||
| XM_011527216.2 | XP_011525518.1 | 4 - 5 | 547 | 88 | ||
| GenBank mRNA | AK057404.1 | - | 4 - 5 | 492 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011527214.1, XM_011527215.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
male meiosis I
spermatid nucleus elongation
sex differentiation
positive regulation of histone H3-K9 dimethylation
positive regulation of histone H3-K9 trimethylation
core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
metal ion binding
transcription, DNA-templated
regulation of transcription, DNA-templated
male meiosis I
spermatid nucleus elongation
sex differentiation
positive regulation of histone H3-K9 dimethylation
positive regulation of histone H3-K9 trimethylation
core promoter proximal region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000269945.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596660.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000596827.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000599022.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000600017.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000601660.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | ENST00000602098.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008163.hg.1 | NM_001040283.2 | Refseq |

DMRTC2
Genomic Map