Assay Details
| Gene Symbol: SGF29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138414.2 | NP_612423.1 | 3 - 4 | 333 | 61 | |
| XM_017022894.1 | XP_016878383.1 | 3 - 4 | 346 | 61 | ||
| GenBank mRNA | AA132818.1 | - | 3 - 4 | 334 | 61 | |
| AK057008.1 | - | 3 - 4 | 292 | 61 | ||
| BC011981.1 | - | 3 - 4 | 324 | 3863665 | 61 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000303.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16004193.hg.1 | ENST00000317058.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004193.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004193.hg.1 | TC16000303.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600152858.hg.1 | TC1600007348.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000317058.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000564682.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000565945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000567564.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | ENST00000569581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007348.hg.1 | NM_138414.2 | Refseq |

SGF29
Genomic Map