Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006582.hg.1 |
ENST00000379800.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006582.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13006582.hg.1 |
TC13000573.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008635.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008635.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008635.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008635.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008635.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008635.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008635.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008635.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008635.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008635.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008635.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008635.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158339.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158339.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158339.hg.1 |
ENST00000379800.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300158339.hg.1 |
NM_145203.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300158339.hg.1 |
TC1300008635.hg.1 |
Clariom D TC |