Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009052.hg.1 |
TC0X000707.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009052.hg.1 |
ENST00000370467.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X009052.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158736.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158736.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158736.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00158736.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00158736.hg.1 |
TC0X00008671.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000370467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
ENST00000489034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00008671.hg.1 |
NM_152578.2 |
Refseq |