Assay Details
| Gene Symbol: RGL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153615.1 | NP_705843.1 | 8 - 9 | 2409 | 74 | |
| XM_017028767.1 | XP_016884256.1 | 2 - 3 | 353 | 74 | ||
| GenBank mRNA | AK131340.1 | - | 11 - 12 | 3014 | 74 | |
| AY101396.1 | - | 8 - 9 | 2409 | 74 | ||
| BC101108.3 | - | 7 - 8 | 2089 | 40011557 | 74 | |
| BC101109.2 | - | 7 - 8 | 2089 | 40011564 | 74 | |
| BC101110.3 | - | 8 - 9 | 1964 | 74 | ||
| HY101412.1 | - | 3 - 4 | 250 | 74 | ||
More Information
Set Membership: |
|

RGL4
Genomic Map