Assay Details
Gene Symbol: ALDH8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022568.3 | NP_072090.1 | 4 - 5 | 665 | 71 | |
NM_170771.2 | NP_739577.1 | 4 - 5 | 665 | 71 | ||
GenBank mRNA | AF303134.1 | - | 4 - 5 | 656 | 71 | |
AK074266.1 | - | 4 - 5 | 658 | 71 | ||
AK222848.1 | - | 4 - 5 | 658 | 71 | ||
AK290784.1 | - | 4 - 5 | 612 | 71 | ||
BC113862.1 | - | 4 - 5 | 635 | 40033493 | 71 | |
BC114473.1 | - | 5 - 6 | 673 | 40033488 | 71 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002132.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000460753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000460753.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000534012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000534012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | ENST00000534012.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | TC06002132.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | ENST00000265605.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | ENST00000367845.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06026786.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600191527.hg.1 | TC0600013249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000265605.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000349305.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000367847.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000460753.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000460753.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000534012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000534012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | ENST00000534012.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_001193480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_022568.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013249.hg.1 | NM_170771.2 | Refseq |
