Assay Details
| Gene Symbol: PPP1R11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK310593.1 | - | 1 - 2 | 328 | 102 | |
| BC003144.1 | - | 1 - 2 | 248 | 2823116 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC003144.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150776.hg.1 | TC0600007508.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376763.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376765.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376769.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376772.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | ENST00000376773.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007508.hg.1 | NM_021959.2 | Refseq |

PPP1R11
Genomic Map