Assay Details
Gene Symbol: KCNQ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172106.2 | NP_742104.1 | 9 - 10 | 1366 | 112 | |
NM_172107.3 | NP_742105.1 | 9 - 10 | 1366 | 112 | ||
NM_172108.4 | NP_742106.1 | 9 - 10 | 1366 | 112 | ||
XM_011528810.1 | XP_011527112.1 | 9 - 10 | 1278 | 112 | ||
XM_017027841.1 | XP_016883330.1 | 9 - 10 | 1278 | 112 | ||
XM_017027842.1 | XP_016883331.1 | 9 - 10 | 1278 | 112 | ||
XM_017027843.1 | XP_016883332.1 | 9 - 10 | 1171 | 112 | ||
XM_017027844.1 | XP_016883333.1 | 9 - 10 | 1278 | 112 | ||
GenBank mRNA | AF033348.1 | - | 9 - 10 | 1279 | 112 | |
AF074247.1 | - | 9 - 10 | 1327 | 112 | ||
AF110020.1 | - | 9 - 10 | 1198 | 112 | ||
AK293727.1 | - | 9 - 10 | 1278 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
