Assay Details
| Gene Symbol: KCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004980.4 | NP_004971.2 | 1 - 2 | 409 | 60 | |
| NM_172198.2 | NP_751948.1 | 1 - 2 | 409 | 60 | ||
| GenBank mRNA | AF120491.1 | - | 1 - 2 | 417 | 60 | |
| AY863198.1 | - | 1 - 2 | 302 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002992.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF120491.1,AY863198.1,NM_004980.4,NM_172198.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
regulation of ion transmembrane transport
protein homooligomerization
cardiac conduction
potassium ion export
cellular response to BMP stimulus
membrane repolarization
A-type (transient outward) potassium channel activity
delayed rectifier potassium channel activity
protein binding
ion channel binding
metal ion binding
potassium ion transport
regulation of ion transmembrane transport
protein homooligomerization
cardiac conduction
potassium ion export
cellular response to BMP stimulus
membrane repolarization
A-type (transient outward) potassium channel activity
delayed rectifier potassium channel activity
protein binding
ion channel binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01046439.hg.1 | TC01002992.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046439.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046439.hg.1 | ENST00000315987.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046439.hg.1 | ENST00000369697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046439.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046439.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046440.hg.1 | TC01002992.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046440.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046440.hg.1 | ENST00000315987.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046440.hg.1 | ENST00000369697.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046440.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046440.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046442.hg.1 | TC01002992.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046442.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046442.hg.1 | ENST00000315987.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046442.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01046442.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214014.hg.1 | TC0100015288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214018.hg.1 | TC0100015288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214012.hg.1 | TC0100015288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214017.hg.1 | TC0100015288.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100015288.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | ENST00000302127.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | ENST00000315987.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | ENST00000369697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | NM_004980.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | NM_172198.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100214011.hg.1 | TC0100015288.hg.1 | Clariom D TC |

KCND3
Genomic Map