Assay Details
Gene Symbol: KRT78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300814.1 | NP_001287743.1 | 2 - 3 | 286 | 79 | |
NM_173352.3 | NP_775487.2 | 2 - 3 | 668 | 79 | ||
XM_011538010.1 | XP_011536312.1 | 2 - 3 | 668 | 79 | ||
GenBank mRNA | AK096419.1 | - | 2 - 3 | 668 | 79 | |
AK290901.1 | - | 2 - 3 | 668 | 79 | ||
AK298636.1 | - | 3 - 4 | 594 | 79 | ||
BX647095.1 | - | 2 - 3 | 286 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK298636.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
