Assay Details
| Gene Symbol: KRT78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300814.1 | NP_001287743.1 | 5 - 6 | 600 | 98 | |
| NM_173352.3 | NP_775487.2 | 5 - 6 | 982 | 98 | ||
| XM_011538010.1 | XP_011536312.1 | 4 - 5 | 886 | 98 | ||
| GenBank mRNA | AK096419.1 | - | 5 - 6 | 982 | 98 | |
| AK290901.1 | - | 5 - 6 | 982 | 98 | ||
| AK298636.1 | - | 6 - 7 | 98 | |||
| BX647095.1 | - | 6 - 7 | 600 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001538.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12020207.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020207.hg.1 | ENST00000359499.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020207.hg.1 | ENST00000539860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020207.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12020207.hg.1 | TC12001538.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000547920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | ENST00000551581.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200010778.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182252.hg.1 | TC1200010778.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | ENST00000304620.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | ENST00000359499.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | NM_001300814.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | NM_173352.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200182253.hg.1 | TC1200010778.hg.1 | Clariom D TC |

KRT78
Genomic Map