Assay Details
Gene Symbol: ANKRD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173595.3 | NP_775866.2 | 22 - 23 | 2582 | 59 | |
XM_011538197.2 | XP_011536499.1 | 21 - 22 | 2684 | 59 | ||
XM_017019183.1 | XP_016874672.1 | 21 - 22 | 2765 | 59 | ||
XM_017019184.1 | XP_016874673.1 | 22 - 23 | 2614 | 59 | ||
GenBank mRNA | AB190990.1 | - | 22 - 23 | 2599 | 59 | |
AK125379.1 | - | 22 - 23 | 2599 | 59 | ||
AK126893.1 | - | 15 - 16 | 1742 | 59 | ||
BC042070.1 | - | 2 - 3 | 208 | 59 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001599.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR12021264.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021264.hg.1 | ENST00000417002.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021264.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12021264.hg.1 | TC12001599.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12021263.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021263.hg.1 | ENST00000417002.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12021263.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12021263.hg.1 | TC12001599.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200183990.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200183990.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200183990.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200183990.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200183990.hg.1 | TC1200010902.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1200183989.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200183989.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200183989.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200183989.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200183989.hg.1 | TC1200010902.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000267116.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000548241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | ENST00000551023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010902.hg.1 | NM_173595.3 | Refseq |
