Assay Details
Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099439.1 | NP_001092909.1 | 2 - 3 | 259 | 104 | |
NM_173641.2 | NP_775912.2 | 2 - 3 | 259 | 104 | ||
GenBank mRNA | AJ781169.1 | - | 2 - 3 | 259 | 104 | |
AK090974.1 | - | 2 - 3 | 240 | 104 | ||
BC067734.1 | - | 2 - 3 | 259 | 4375803 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
