Assay Details
| Gene Symbol: FER1L6-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040044.1 | 4 - 5 | 636 | 82 | ||
| GenBank mRNA | AK093040.1 | - | 4 - 5 | 636 | 82 | |
| BC137357.1 | - | 4 - 5 | 603 | 9021355 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001595.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P322013 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 17484 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 57912 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P322013 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P322013 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8152740 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8152737 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8152741 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8152736 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8152735 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8152738 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8152739 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8152734 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1553443_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020870.hg.1 | TC08001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020870.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020870.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020869.hg.1 | TC08001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020869.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020869.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020868.hg.1 | TC08001595.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020868.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08020868.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179422.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179422.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179422.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179422.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179422.hg.1 | TC0800011697.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800011697.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179423.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179423.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179423.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179423.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179423.hg.1 | TC0800011697.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800179424.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179424.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179424.hg.1 | ENST00000518567.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800179424.hg.1 | NR_040044.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800179424.hg.1 | TC0800011697.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1553443_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1553443_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1553443_at3 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036112 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 218925 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1788415 | NR_040044.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1788415 | AK093040.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1788415 | NR_040044.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1788415 | AK093040.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1788415 | NR_040044.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1788415 | AK093040.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1788415 | NR_040044.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1788415 | AK093040.1 | human_transcriptome |

FER1L6-AS1
Genomic Map