Assay Details
| Gene Symbol: HIST2H2AC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003517.2 | NP_003508.1 | 1 | 381 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006326.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501652 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 148986 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 3639 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P301247 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P8721 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P301247 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 8476 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 23800 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P101364 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P101364 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 25252 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 61816 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P301247 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P301247 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501652 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P301247 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P8721 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P101364 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501653 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00501653 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P101364 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P101364 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P301247 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P8721 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 6299 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 1949 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 20481 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P101364 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501652 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501682 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501650 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501690 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501673 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P101364 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_003517_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905081 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919623 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905089 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905091 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7905090 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7919619 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7905088 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7905079 | - | - |
| Affymetrix Human Genome U219 Array | 11715204_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11759258_at | - | - |
| Affymetrix Human Genome U219 Array | 11715204_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11759258_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018007.hg.1 | TC01006326.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018007.hg.1 | ENST00000331380.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018007.hg.1 | NM_003517.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171265.hg.1 | NM_003517.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171265.hg.1 | NM_003517.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171265.hg.1 | TC0100009880.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030494 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A030494 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_27436923-S | BC060324.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_27436923-S | NM_003517.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_13493-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0003360408 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0003360408 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 5860075 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_13493 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1768973 | NM_003517.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768973 | BC060324.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1768973 | NM_003517.2 | human_transcriptome |

HIST2H2AC
Genomic Map