Assay Details
| Gene Symbol: KCNH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139318.4 | NP_647479.2 | 2 - 3 | 243 | 84 | |
| NM_172375.2 | NP_758963.1 | 2 - 3 | 243 | 84 | ||
| GenBank mRNA | AF418206.1 | - | 2 - 3 | 191 | 84 | |
| AF472412.1 | - | 2 - 3 | 191 | 84 | ||
| AF493798.1 | - | 2 - 3 | 191 | 84 | ||
| AK074484.1 | - | 2 - 3 | 185 | 84 | ||
| BC043409.2 | - | 2 - 3 | 360 | 84 | ||
| BC073979.1 | - | 2 - 3 | 243 | 6199697 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001202.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK074484.1,BC043409.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
regulation of G2/M transition of mitotic cell cycle
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
phosphorelay sensor kinase activity
voltage-gated potassium channel activity
calmodulin binding
ion channel binding
protein heterodimerization activity
phosphorelay signal transduction system
regulation of G2/M transition of mitotic cell cycle
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of membrane potential
potassium ion transmembrane transport
phosphorelay sensor kinase activity
voltage-gated potassium channel activity
calmodulin binding
ion channel binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | ENST00000322893.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | ENST00000394964.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | ENST00000394968.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | ENST00000420622.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14013723.hg.1 | TC14001202.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009381.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000322893.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000394964.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000394968.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | ENST00000420622.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | NM_139318.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | NM_172375.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400169309.hg.1 | TC1400009381.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 103370 | - | - |

KCNH5
Genomic Map