Assay Details
| Gene Symbol: CAPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000070.2 | NP_000061.1 | 16 - 17 | 2219 | 73 | |
| NM_024344.1 | NP_077320.1 | 15 - 16 | 2201 | 73 | ||
| NM_173088.1 | NP_775111.1 | 5 - 6 | 546 | 73 | ||
| GenBank mRNA | AF127764.1 | - | 15 - 16 | 2116 | 73 | |
| AK308103.1 | - | 16 - 17 | 2219 | 73 | ||
| BC003169.1 | - | 5 - 6 | 546 | 73 | ||
| BC107791.1 | - | 2 - 3 | 309 | 73 | ||
| BC146649.1 | - | 16 - 17 | 2132 | 40147158 | 73 | |
| BC146672.1 | - | 16 - 17 | 2132 | 40148444 | 73 | |
| BT007322.1 | - | 4 - 5 | 377 | 73 | ||
| EU791850.1 | - | 16 - 17 | 1652 | 73 | ||
| EU791851.1 | - | 15 - 16 | 1508 | 73 | ||
| KU177954.1 | - | 4 - 5 | 377 | 73 | ||
| KU177955.1 | - | 4 - 5 | 478 | 73 | ||
| X85030.1 | - | 16 - 17 | 1913 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC107791.1 transcripts.
Set Membership: |
|

CAPN3
Genomic Map