Assay Details
| Gene Symbol: TSC22D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303264.1 | NP_001290193.1 | 1 - 2 | 2974 | 99 | |
| GenBank mRNA | AF201291.1 | - | 1 - 2 | 101 | 99 | |
| AK297383.1 | - | 2 - 3 | 868 | 99 | ||
| AK297748.1 | - | 1 - 2 | 303 | 99 | ||
More Information
Set Membership: |
|

TSC22D2
Genomic Map