Assay Details
Gene Symbol: PRODH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021232.1 | NP_067055.1 | 9 - 10 | 1358 | 98 | |
XM_017027084.1 | XP_016882573.1 | 8 - 9 | 1215 | 98 | ||
GenBank mRNA | BC039710.1 | - | 8 - 9 | 1130 | 98 | |
U80018.1 | - | 9 - 10 | 1358 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19001455.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019658.hg.1 | ENST00000301175.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19019658.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19019658.hg.1 | TC19001455.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000588266.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | ENST00000591694.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900010499.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | NM_021232.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | TC1900010499.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000301175.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000587695.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000587808.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | ENST00000589835.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900185945.hg.1 | NM_021232.1 | Refseq |
