Assay Details
| Gene Symbol: HIST2H2AB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175065.2 | NP_778235.1 | 1 | 383 | 153 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003194.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558072 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501679 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558078 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501655 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501677 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501661 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501678 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558068 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501654 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501662 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558062 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501671 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558084 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501666 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501667 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501659 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501669 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P20558076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P00501660 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 63400 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 62111 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 107017 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P68631 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P343927 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P68631 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 44474 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 15217 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P130835 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P130835 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 61303 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P68631 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P68631 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501666 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501660 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501662 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558078 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558068 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558079 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558076 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501680 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501669 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501659 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501671 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501679 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558072 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558075 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501658 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558083 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501678 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501670 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558062 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20558084 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501655 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00501666 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P343927 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P68631 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P130835 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501685 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501658 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501670 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501672 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501664 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501679 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501656 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501683 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501682 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501690 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501676 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501688 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501668 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501655 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501666 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501662 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501680 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P00501659 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00501654 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00501655 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P00501656 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P130835 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P130835 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P343927 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P68631 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 1653 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 6788 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 4104 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 56313 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P130835 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501617 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501656 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501640 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501645 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501679 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501685 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501658 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501688 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501612 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501631 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501682 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501622 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501635 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501673 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501668 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501624 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501680 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501659 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501636 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501629 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501625 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501666 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501627 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501633 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501647 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15126929 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501655 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501664 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501614 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501677 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501637 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501670 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501621 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501639 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501662 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501619 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501616 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501642 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00501690 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P130835 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_175065_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919643 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7919644 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7919642 | - | - |
| Affymetrix Human Genome U219 Array | 11715149_PM_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11759284_at | - | - |
| Affymetrix Human Genome U219 Array | 11715149_x_at | - | - |
| Affymetrix Human Genome U219 Array | 11715148_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11715148_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11759284_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048931.hg.1 | TC01003194.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048931.hg.1 | ENST00000331128.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01048931.hg.1 | NM_175065.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217612.hg.1 | NM_175065.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217612.hg.1 | NM_175065.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100217612.hg.1 | TC0100015711.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051656 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A051656 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 172307 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_29171728-S | NM_175065.2 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_10158-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0001570441 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0001570441 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2470243 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_10158 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1747589 | NM_175065.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1747589 | NM_175065.2 | human_transcriptome |

HIST2H2AB
Genomic Map