Assay Details
| Gene Symbol: EPB41L4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018424.3 | NP_060894.2 | 13 - 14 | 1826 | 131 | |
| NM_019114.4 | NP_061987.3 | 13 - 14 | 1826 | 131 | ||
| XM_011518790.1 | XP_011517092.1 | 13 - 14 | 1323 | 131 | ||
| XM_011518791.2 | XP_011517093.1 | 13 - 14 | 1308 | 131 | ||
| XM_011518792.2 | XP_011517094.1 | 13 - 14 | 1308 | 131 | ||
| XM_011518794.2 | XP_011517096.1 | 13 - 14 | 1308 | 131 | ||
| XM_017014813.1 | XP_016870302.1 | 13 - 14 | 1308 | 131 | ||
| XM_017014814.1 | XP_016870303.1 | 13 - 14 | 1308 | 131 | ||
| XM_017014815.1 | XP_016870304.1 | 13 - 14 | 1308 | 131 | ||
| GenBank mRNA | AB032179.3 | - | 13 - 14 | 1826 | 131 | |
| AB209187.1 | - | 5 - 6 | 418 | 131 | ||
| AF153416.1 | - | 13 - 14 | 1598 | 131 | ||
| AF153418.1 | - | 13 - 14 | 1598 | 131 | ||
| AK000388.1 | - | 13 - 14 | 1210 | 131 | ||
More Information
Set Membership: |
|

EPB41L4B
Genomic Map