Assay Details
| Gene Symbol: PLA2G4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178034.3 | NP_828848.3 | 16 - 17 | 1776 | 138 | |
| XM_011521467.2 | XP_011519769.1 | 15 - 16 | 1607 | 138 | ||
| GenBank mRNA | AB090876.1 | - | 16 - 17 | 1779 | 138 | |
| BC034571.1 | - | 2 - 3 | 213 | 138 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001259.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phospholipid metabolic process
phosphatidic acid biosynthetic process
phospholipid catabolic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
lysophospholipase activity
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
metal ion binding
phospholipid metabolic process
phosphatidic acid biosynthetic process
phospholipid catabolic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylinositol acyl-chain remodeling
phosphatidylserine acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
lysophospholipase activity
phospholipase A2 activity
phosphatidylcholine 1-acylhydrolase activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15011515.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011515.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15011515.hg.1 | TC15001259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500166564.hg.1 | TC1500009179.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000290472.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560132.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | ENST00000560932.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009179.hg.1 | NM_178034.3 | Refseq |

PLA2G4D
Genomic Map