Assay Details
| Gene Symbol: NT5DC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152729.2 | NP_689942.2 | 4 - 5 | 459 | 71 | |
| XM_006715378.3 | XP_006715441.1 | 4 - 5 | 462 | 71 | ||
| GenBank mRNA | AK090918.1 | - | 1 - 2 | 1189 | 71 | |
| AK290408.1 | - | 4 - 5 | 409 | 71 | ||
| AK308337.1 | - | 4 - 5 | 442 | 71 | ||
| AY203935.1 | - | 1 - 2 | 69 | 71 | ||
| BC015138.1 | - | 3 - 4 | 240 | 71 | ||
| BC108699.1 | - | 4 - 5 | 416 | 6061738 | 71 | |
| BC136832.1 | - | 4 - 5 | 418 | 9020823 | 71 | |
| BC136841.1 | - | 4 - 5 | 418 | 9020832 | 71 | |
| BM808222.1 | - | 448 | 71 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000916.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY203935.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06010691.hg.1 | TC06000916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010691.hg.1 | ENST00000319550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010691.hg.1 | ENST00000368618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010691.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010689.hg.1 | TC06000916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010689.hg.1 | ENST00000319550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010689.hg.1 | ENST00000368618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010689.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010694.hg.1 | TC06000916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010694.hg.1 | ENST00000319550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010694.hg.1 | ENST00000368618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010694.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010687.hg.1 | TC06000916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010687.hg.1 | ENST00000319550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010687.hg.1 | ENST00000368618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010687.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010690.hg.1 | TC06000916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010690.hg.1 | ENST00000319550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010690.hg.1 | ENST00000368618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010690.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010695.hg.1 | TC06000916.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010695.hg.1 | ENST00000319550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010695.hg.1 | ENST00000368618.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06010695.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008469.hg.1 | ENST00000399325.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163317.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163321.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163325.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | ENST00000460749.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600009228.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163320.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | ENST00000417846.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163326.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008469.hg.1 | ENST00000399325.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163319.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | ENST00000319550.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | ENST00000419791.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | NM_152729.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600163322.hg.1 | TC0600009228.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 118048 | - | - |

NT5DC1
Genomic Map