Assay Details
| Gene Symbol: IGDCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004884.3 | NP_004875.2 | 12 - 13 | 2251 | 62 | |
| XM_011522241.1 | XP_011520543.1 | 12 - 13 | 2193 | 62 | ||
| XM_011522243.1 | XP_011520545.1 | 11 - 12 | 1777 | 62 | ||
| XM_011522244.1 | XP_011520546.1 | 11 - 12 | 1766 | 62 | ||
| GenBank mRNA | AF063936.1 | - | 4 - 5 | 483 | 62 | |
| BC042054.1 | - | 12 - 13 | 2199 | 5579570 | 62 | |
| BC067107.1 | - | 12 - 13 | 2251 | 6644528 | 62 | |
| BX389424.1 | - | 2 - 3 | 188 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001565.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | ENST00000559947.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014941.hg.1 | ENST00000327987.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014941.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014941.hg.1 | TC15001565.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000558354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | ENST00000559947.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009795.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172382.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172382.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172382.hg.1 | ENST00000327987.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172382.hg.1 | NM_004884.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172382.hg.1 | TC1500009795.hg.1 | Clariom D TC |

IGDCC3
Genomic Map