Assay Details
| Gene Symbol: RPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000999.3 | NP_000990.1 | 1 - 2 | 86 | 115 | |
| GenBank mRNA | AK097905.1 | - | 1 - 2 | 80 | 286 | |
| AK312119.1 | - | 1 - 2 | 54 | 115 | ||
| CB124619.1 | - | 1 - 2 | 76 | 115 | ||
| Z26876.1 | - | 1 - 2 | 68 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000825.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312119.1,NM_000999.3,Z26876.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
skeletal system development
ossification
rRNA processing
translation
translational initiation
regulation of translation
SRP-dependent cotranslational protein targeting to membrane
sensory perception of sound
viral transcription
ribonucleoprotein complex assembly
90S preribosome assembly
middle ear morphogenesis
axial mesoderm development
RNA binding
structural constituent of ribosome
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
skeletal system development
ossification
rRNA processing
translation
translational initiation
regulation of translation
SRP-dependent cotranslational protein targeting to membrane
sensory perception of sound
viral transcription
ribonucleoprotein complex assembly
90S preribosome assembly
middle ear morphogenesis
axial mesoderm development
RNA binding
structural constituent of ribosome
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17010619.hg.1 | ENST00000311111.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010619.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010619.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010619.hg.1 | TC17000825.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000439590.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008842.hg.1 | NM_001035258.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000311111.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000528433.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000529123.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000530646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000533498.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000534490.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | ENST00000584577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | NM_000999.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700165944.hg.1 | TC1700008842.hg.1 | Clariom D TC |

RPL38
Genomic Map