Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008683.hg.1 |
ENST00000610599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008683.hg.1 |
ENST00000610599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008683.hg.1 |
ENST00000610599.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009780.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009780.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009780.hg.1 |
TC20000720.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009778.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009778.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009778.hg.1 |
TC20000720.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009779.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009779.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20009779.hg.1 |
TC20000720.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160800.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160800.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160800.hg.1 |
TC2000008686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160800.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160800.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008686.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160797.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160797.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160797.hg.1 |
TC2000008686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160797.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160797.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160798.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160798.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000160798.hg.1 |
TC2000008686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000160798.hg.1 |
ENST00000304710.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000160798.hg.1 |
NM_001900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008683.hg.1 |
ENST00000610599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008683.hg.1 |
ENST00000610599.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008683.hg.1 |
ENST00000610599.1 |
Ensembl |