Assay Details
| Gene Symbol: HIST1H2BO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003527.4 | NP_003518.2 | 1 | 56 | 81 | |
| GenBank mRNA | BC106720.2 | - | 1 | 39 | 40014271 | 81 |
| BX647290.1 | - | 1 | 56 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000270.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC106720.2,NM_003527.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630377 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820583 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630390 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630385 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820602 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630406 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820618 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820612 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820613 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820596 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820620 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820603 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820593 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820607 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820617 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630400 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630396 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630380 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820606 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820600 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820614 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820608 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630413 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820592 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630402 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820601 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630407 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820622 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630383 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P24820610 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P04630391 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 77501 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 35386 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P59069 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P59069 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 16999 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P122601 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P122601 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 23290 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P59069 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00316450 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P59069 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630383 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820621 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820603 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630400 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630389 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820607 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630390 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630377 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630396 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820603 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630379 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630396 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820612 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630406 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820618 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630390 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630380 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630400 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820592 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820600 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820621 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820601 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820614 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820613 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630377 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630389 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820617 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820607 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820596 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24820602 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04630383 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P59069 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P122601 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630399 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630401 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630395 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630406 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630381 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630383 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630390 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630396 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630403 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630408 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630385 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630389 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630379 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630394 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630392 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630405 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P04630377 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630376 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04630375 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P122601 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P122601 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P59069 | - | - |
| Custom Human Agilent Array 11k, Chromosome 6, Expression | 5186 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 155199 | AK098366.1 | genbank_mrna_human |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 155199 | BC045760.1 | genbank_mrna_human |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 68863 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P122601 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630394 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630399 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630396 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630405 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630374 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630409 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630398 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630413 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630411 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630392 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630383 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630403 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630385 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630415 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630379 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630401 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P04630389 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P122601 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_003527_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117617 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117618 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117615 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8117616 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8117614 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 214540_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742176_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11742176_PM_s_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002259.hg.1 | TC06000270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002259.hg.1 | ENST00000303806.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002259.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002260.hg.1 | TC06000270.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002260.hg.1 | ENST00000303806.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06002260.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007384.hg.1 | ENST00000616182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007384.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007384.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007384.hg.1 | ENST00000616182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007384.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007384.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150046.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150046.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150046.hg.1 | TC0600007384.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600150045.hg.1 | ENST00000616182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150045.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150045.hg.1 | ENST00000616182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600150045.hg.1 | NM_003527.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600150045.hg.1 | TC0600007384.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 214540_at | - | - |
| Human Genome U133 Plus 2.0 Array | 214540_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214540_PM_at | - | - |
| Human Genome U133A 2.0 Array | 214540_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003527_at | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw503599 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 214540_at11 | - | - |
| Human Genome U133 Set | 214540_PM_at | - | - |
| Human Genome U133 Set | 214540_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031158 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A031158 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 105346 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_16306565-S | NM_003527.4 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_18509-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0004480280 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0004480280 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_18509 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_18509 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1739076 | NM_003527.4 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1739076 | NM_003527.4 | human_transcriptome |

HIST1H2BO
Genomic Map