Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002689.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002689.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002689.hg.1 |
TC20000216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009868.hg.1 |
ENST00000435438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002687.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002687.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002687.hg.1 |
TC20000216.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002690.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002690.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002690.hg.1 |
TC20000216.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000148789.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148789.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148789.hg.1 |
TC2000007133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148789.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148789.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148793.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148793.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148793.hg.1 |
TC2000007133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148793.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148793.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009868.hg.1 |
ENST00000435438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148791.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148791.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148791.hg.1 |
TC2000007133.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148791.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148791.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
ENST00000375571.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007133.hg.1 |
NM_012325.2 |
Refseq |