Assay Details
| Gene Symbol: KCNH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002238.3 | NP_002229.1 | 3 - 4 | 479 | 124 | |
| NM_172362.2 | NP_758872.1 | 3 - 4 | 479 | 124 | ||
| GenBank mRNA | AF078741.1 | - | 3 - 4 | 345 | 124 | |
| AF078742.1 | - | 3 - 4 | 345 | 124 | ||
| AJ001366.1 | - | 3 - 4 | 479 | 124 | ||
| BC113709.1 | - | 3 - 4 | 379 | 8322761 | 124 | |
| BC143599.1 | - | 3 - 4 | 380 | 9052111 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003791.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphorelay signal transduction system
startle response
potassium ion transport
myoblast fusion
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of cell proliferation
regulation of membrane potential
cellular response to calcium ion
potassium ion transmembrane transport
phosphorelay sensor kinase activity
delayed rectifier potassium channel activity
protein binding
calmodulin binding
protein kinase binding
cyclic nucleotide binding
identical protein binding
ion channel binding
protein heterodimerization activity
14-3-3 protein binding
phosphorelay signal transduction system
startle response
potassium ion transport
myoblast fusion
signal transduction by protein phosphorylation
regulation of ion transmembrane transport
regulation of cell proliferation
regulation of membrane potential
cellular response to calcium ion
potassium ion transmembrane transport
phosphorelay sensor kinase activity
delayed rectifier potassium channel activity
protein binding
calmodulin binding
protein kinase binding
cyclic nucleotide binding
identical protein binding
ion channel binding
protein heterodimerization activity
14-3-3 protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01058046.hg.1 | TC01003791.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058046.hg.1 | ENST00000271751.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058046.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058046.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01058046.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100230290.hg.1 | TC0100017193.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000271751.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | ENST00000367007.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_002238.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017193.hg.1 | NM_172362.2 | Refseq |

KCNH1
Genomic Map