Assay Details
Gene Symbol: LMBRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018368.3 | NP_060838.3 | 11 - 12 | 1357 | 134 | |
XM_011535941.1 | XP_011534243.1 | 11 - 12 | 1207 | 134 | ||
GenBank mRNA | AF113224.1 | - | 11 - 12 | 1077 | 134 | |
AF208863.1 | - | 10 - 11 | 1205 | 134 | ||
AF211480.1 | - | 8 - 9 | 695 | 134 | ||
AK002102.1 | - | 11 - 12 | 1106 | 134 | ||
AK225187.1 | - | 11 - 12 | 1106 | 134 | ||
AK225537.1 | - | 11 - 12 | 1091 | 134 | ||
AK290069.1 | - | 11 - 12 | 1237 | 134 | ||
AY136817.1 | - | 10 - 11 | 862 | 134 | ||
BC047073.1 | - | 11 - 12 | 1196 | 5274221 | 134 | |
BX647737.1 | - | 10 - 11 | 949 | 134 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001854.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cobalamin metabolic process
cobalamin transport
viral process
insulin receptor internalization
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of protein kinase B signaling
insulin receptor binding
cobalamin transporter activity
cobalamin binding
cobalamin metabolic process
cobalamin transport
viral process
insulin receptor internalization
negative regulation of glucose import
negative regulation of insulin receptor signaling pathway
negative regulation of protein kinase B signaling
insulin receptor binding
cobalamin transporter activity
cobalamin binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06022487.hg.1 | TC06001854.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06022487.hg.1 | ENST00000370570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022487.hg.1 | ENST00000370577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022487.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06022489.hg.1 | TC06001854.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06022489.hg.1 | ENST00000370570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022489.hg.1 | ENST00000370577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022489.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06022488.hg.1 | TC06001854.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06022488.hg.1 | ENST00000370570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022488.hg.1 | ENST00000370577.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06022488.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184236.hg.1 | TC0600012240.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184237.hg.1 | TC0600012240.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600184238.hg.1 | TC0600012240.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370570.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000370577.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | ENST00000472827.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600012240.hg.1 | NM_018368.3 | Refseq |
