Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016800.hg.1 |
TC09001339.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016800.hg.1 |
ENST00000287996.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016800.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016801.hg.1 |
TC09001339.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016801.hg.1 |
ENST00000287996.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09016801.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000375522.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
ENST00000486841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900010804.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173564.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173564.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173564.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173564.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173564.hg.1 |
TC0900010804.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900173565.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173565.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173565.hg.1 |
ENST00000287996.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900173565.hg.1 |
NM_022755.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900173565.hg.1 |
TC0900010804.hg.1 |
Clariom D TC |