Assay Details
| Gene Symbol: ENPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153343.3 | NP_699174.1 | 1 - 2 | 382 | 75 | |
| GenBank mRNA | AK057370.1 | - | 1 - 2 | 327 | 75 | |
| AY358676.1 | - | 1 - 2 | 323 | 75 | ||
| BC035035.2 | - | 1 - 2 | 287 | 5259487 | 75 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001800.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04024547.hg.1 | TC04001800.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024547.hg.1 | ENST00000296741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024547.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024546.hg.1 | TC04001800.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024546.hg.1 | ENST00000296741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024546.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024543.hg.1 | TC04001800.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024543.hg.1 | ENST00000296741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024543.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024548.hg.1 | TC04001800.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024548.hg.1 | ENST00000296741.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04024548.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186173.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186173.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186173.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186173.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186173.hg.1 | TC0400012595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186175.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186175.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186175.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186175.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186175.hg.1 | TC0400012595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000510054.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012595.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | ENST00000512353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186169.hg.1 | TC0400012595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | ENST00000505644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186172.hg.1 | TC0400012595.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400186174.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186174.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186174.hg.1 | ENST00000296741.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400186174.hg.1 | NM_153343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400186174.hg.1 | TC0400012595.hg.1 | Clariom D TC |

ENPP6
Genomic Map