Assay Details
| Gene Symbol: GLDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000170.2 | NP_000161.2 | 11 - 12 | 1675 | 69 | |
| GenBank mRNA | AK310534.1 | - | 7 - 8 | 1050 | 69 | |
| AK314156.1 | - | 11 - 12 | 1677 | 69 | ||
| BC111993.1 | - | 11 - 12 | 1548 | 8327461 | 69 | |
| BC111995.1 | - | 11 - 12 | 1548 | 8327463 | 69 | |
| D90239.1 | - | 11 - 12 | 1632 | 69 | ||
| M63635.1 | - | 11 - 12 | 1563 | 69 | ||
| M64590.1 | - | 11 - 12 | 1632 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000899.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09011949.hg.1 | TC09000899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011949.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09011949.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900165100.hg.1 | TC0900009489.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000321612.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000460457.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000463305.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000467946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | ENST00000477960.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009489.hg.1 | NM_000170.2 | Refseq |

GLDC
Genomic Map