Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008816.hg.1 |
ENST00000268035.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008816.hg.1 |
ENST00000558762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008816.hg.1 |
NM_000875.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15008816.hg.1 |
TC15000949.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000268035.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000558762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000558898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000559582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000559925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
NM_000875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
NM_001291858.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000268035.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000558762.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000558898.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000559582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
ENST00000559925.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
NM_000875.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
NM_001291858.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500161524.hg.1 |
TC1500008485.hg.1 |
Clariom D TC |