| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021026.hg.1 |
TC09001653.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021026.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021026.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021026.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021024.hg.1 |
TC09001653.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021024.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021024.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021024.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021027.hg.1 |
TC09001653.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021027.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09021027.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180088.hg.1 |
TC0900011708.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180091.hg.1 |
TC0900011708.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0900180093.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180093.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180093.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900180093.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900180093.hg.1 |
TC0900011708.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000340607.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
ENST00000481476.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900011708.hg.1 |
NM_004878.4 |
Refseq |