Assay Details
| Gene Symbol: NR5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004959.4 | NP_004950.2 | 1 - 2 | 171 | 71 | |
| GenBank mRNA | BC032501.1 | - | 1 - 2 | 171 | 5227690 | 71 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001579.hg.1
Additional Information:
This assay is in the 5'UTR region of the BC032501.1,NM_004959.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
luteinization
transcription initiation from RNA polymerase II promoter
cell-cell signaling
primary sex determination
male gonad development
hormone-mediated signaling pathway
tissue development
multicellular organism aging
cell differentiation
adrenal gland development
intracellular receptor signaling pathway
hormone metabolic process
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
regulation of steroid biosynthetic process
maintenance of protein location in nucleus
positive regulation of male gonad development
negative regulation of female gonad development
transcription regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
phospholipid binding
zinc ion binding
enzyme binding
luteinization
transcription initiation from RNA polymerase II promoter
cell-cell signaling
primary sex determination
male gonad development
hormone-mediated signaling pathway
tissue development
multicellular organism aging
cell differentiation
adrenal gland development
intracellular receptor signaling pathway
hormone metabolic process
steroid hormone mediated signaling pathway
positive regulation of transcription from RNA polymerase II promoter
regulation of steroid biosynthetic process
maintenance of protein location in nucleus
positive regulation of male gonad development
negative regulation of female gonad development
transcription regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
steroid hormone receptor activity
transcription coactivator activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
phospholipid binding
zinc ion binding
enzyme binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09019816.hg.1 | TC09001579.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019816.hg.1 | ENST00000373588.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019816.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019814.hg.1 | TC09001579.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019814.hg.1 | ENST00000373588.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09019814.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373587.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011499.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | ENST00000455734.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900178416.hg.1 | TC0900011499.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | ENST00000373588.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | ENST00000620110.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | NM_004959.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900178419.hg.1 | TC0900011499.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 196795 | - | - |

NR5A1
Genomic Map