Assay Details
Gene Symbol: KIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012311.3 | NP_036443.1 | 2 - 3 | 309 | 81 | |
NR_045609.1 | 2 - 3 | 309 | 81 | |||
XM_006717434.3 | XP_006717497.1 | 2 - 3 | 304 | 81 | ||
XM_011519427.1 | XP_011517729.1 | 2 - 3 | 304 | 81 | ||
GenBank mRNA | AJ005273.1 | - | 2 - 3 | 280 | 81 | |
AK301789.1 | - | 2 - 3 | 309 | 81 | ||
AK309059.1 | - | 2 - 3 | 245 | 81 | ||
BC017309.2 | - | 2 - 3 | 226 | 5089578 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001022.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012745.hg.1 | ENST00000379562.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012745.hg.1 | ENST00000535925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012745.hg.1 | ENST00000543003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012745.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012745.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012745.hg.1 | TC10001022.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000460089.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000463666.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000471320.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | ENST00000498098.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009742.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | ENST00000379562.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | NM_012311.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | NR_045609.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000166535.hg.1 | TC1000009742.hg.1 | Clariom D TC |
