Assay Details
| Gene Symbol: CASP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032974.4 | NP_116756.2 | 5 - 6 | 1106 | 111 | |
| NM_032977.3 | NP_116759.2 | 5 - 6 | 1106 | 111 | ||
| GenBank mRNA | AF111345.1 | - | 4 - 5 | 688 | 111 | |
| AK310746.1 | - | 5 - 6 | 892 | 111 | ||
| BC042844.1 | - | 5 - 6 | 842 | 5176698 | 111 | |
| KU177959.1 | - | 4 - 5 | 688 | 111 | ||
| U86214.1 | - | 5 - 6 | 882 | 111 | ||
More Information
Set Membership: |
|

CASP10
Genomic Map