Assay Details
| Gene Symbol: LMF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033200.2 | NP_149977.2 | 3 - 4 | 409 | 61 | |
| XM_005261955.3 | XP_005262012.1 | 3 - 4 | 491 | 61 | ||
| XM_006724426.3 | XP_006724489.1 | 2 - 3 | 940 | 61 | ||
| XM_006724427.3 | XP_006724490.1 | 3 - 4 | 491 | 61 | ||
| XM_017029077.1 | XP_016884566.1 | 3 - 4 | 543 | 61 | ||
| GenBank mRNA | BC002942.2 | - | 3 - 4 | 383 | 3954961 | 61 |
| BC014652.1 | - | 3 - 4 | 472 | 4562676 | 61 | |
| BC021143.1 | - | 3 - 4 | 383 | 4872642 | 61 | |
| BI917400.1 | - | 3 - 4 | 409 | 61 | ||
| CR456605.1 | - | 3 - 4 | 393 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000914.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22015601.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015601.hg.1 | ENST00000380796.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015601.hg.1 | ENST00000474879.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015601.hg.1 | ENST00000505981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015601.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22015601.hg.1 | TC22000914.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | TC2200009161.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200166507.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000216080.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000474879.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000504717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000505981.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000507607.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | ENST00000514938.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009161.hg.1 | NM_033200.2 | Refseq |

LMF2
Genomic Map