Assay Details
Gene Symbol: RAD54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205262.2 | NP_001192191.1 | 3 - 4 | 436 | 68 | |
GenBank mRNA | BC020668.1 | - | 3 - 4 | 403 | 68 | |
BC033710.2 | - | 2 - 3 | 321 | 68 | ||
CN356732.1 | - | 3 - 4 | 416 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC033710.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
