Assay Details
| Gene Symbol: ARL6IP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152522.5 | NP_689735.1 | 2 - 3 | 1177 | 112 | |
| GenBank mRNA | AF530059.1 | - | 2 - 3 | 445 | 112 | |
| AK023109.1 | - | 2 - 3 | 1156 | 112 | ||
| AK315657.1 | - | 2 - 3 | 473 | 112 | ||
| BC028741.2 | - | 2 - 3 | 457 | 5263418 | 112 | |
| BC070140.1 | - | 2 - 3 | 448 | 4672885 | 112 | |
More Information
Set Membership: |
|

ARL6IP6
Genomic Map