Assay Details
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152769.2 | NP_689982.3 | 6 - 7 | 883 | 112 | |
XM_017026555.1 | XP_016882044.1 | 6 - 7 | 1002 | 112 | ||
XM_017026556.1 | XP_016882045.1 | 6 - 7 | 857 | 112 | ||
XM_017026559.1 | XP_016882048.1 | 6 - 7 | 1002 | 112 | ||
GenBank mRNA | BC028156.1 | - | 6 - 7 | 734 | 5241080 | 112 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC19000998.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19013642.hg.1 | ENST00000215376.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19013642.hg.1 | ENST00000382477.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR19013642.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR19013642.hg.1 | TC19000998.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000215376.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1900009137.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | TC1900009137.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172657.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | TC1900009137.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000589260.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172659.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | NM_152769.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | TC1900009137.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | ENST00000382477.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | ENST00000590083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | ENST00000591127.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1900172656.hg.1 | NM_152769.2 | Refseq |
