Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002473.hg.1 |
ENST00000262659.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002473.hg.1 |
ENST00000300415.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002473.hg.1 |
ENST00000339619.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002473.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002473.hg.1 |
TC20000204.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000148491.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148491.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148491.hg.1 |
TC2000007100.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148491.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148491.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000262659.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
ENST00000452892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007100.hg.1 |
NM_080625.3 |
Refseq |